1,2-Difluoro-4-nitrobenzene, 98+%
1,2-Difluoro-4-nitrobenzene, 98+%
1,2-Difluoro-4-nitrobenzene, 98+%
1,2-Difluoro-4-nitrobenzene, 98+%
Thermo Scientific Chemicals

1,2-Difluoro-4-nitrobenzene, 98+%

CAS: 369-34-6 | C6H3F2NO2 | 159.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A11060.18
also known as A11060-18
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS369-34-6
IUPAC Name1,2-difluoro-4-nitrobenzene
Molecular FormulaC6H3F2NO2
InChI KeyRUBQQRMAWLSCCJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive Index1.5060-1.5100 @ 20?C
Assay (GC)≥98.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only