Note: Bac-to-Bac™ Expression System kits now include the NEW! ExpiFectamine Sf Transfection Reagent.
Bac-to-Bac™ N-His TOPO™ Expression System
Gibco™

Bac-to-Bac™ N-His TOPO™ Expression System

Bac-to-Bac baculovirus expression is an efficient method for producing baculovirus for high-level protein expression in insect cells. It relies onRead more
Have Questions?
Catalog NumberQuantity
A1110120 reactions
Catalog number A11101
Price (USD)
4,795.00
Each
Quantity:
20 reactions
Price (USD)
4,795.00
Each
Bac-to-Bac baculovirus expression is an efficient method for producing baculovirus for high-level protein expression in insect cells. It relies on generation of recombinant virus by site-specific transposition in E. coli rather than homologous recombination in insect cells. The pFastBac N-His TOPO expression system features:

• Flexibility—The Bac-to-Bac N-His TOPO vector contains an N-terminal His-tag with a TEV cleavage site to enable easy purification of His-tagged proteins with nickel-chelating resins (such as the Invitrogen ProBond Purification System) and generation of native proteins with the aid of the Invitrogen AcTEV Protease.

• Fast cloning, easy screening—Blunt TOPO cloning technology enables cloning of blunt-end PCR products into the pFastBac TOPO vector in five minutes. In addition, the pFastBac TOPO expression kit is supplied with Mach1-T1R E. coli that enable the visualization of colonies eight hours after plating on ampicillin-selective plates due to their faster doubling time compared to other standard cloning strains. Find out more about the advantages of TOPO cloning ›

• High transfection efficiency with ExpiFectamine Sf Transfection Reagent—The Bac-to-Bac TOPO expression kits now come with the next-generation ExpiFectamine Sf Transfection Reagent for efficient DNA transfection in insect cells using fast, flexible protocols. Find out more about this reagent ›

• Time-saving expression bacmid—With the Bac-to-Bac system, the expression cassette of the pFastBac vector recombines with the parent bacmid in DH10Bac E. coli Competent Cells to form an expression bacmid. The bacmid is then transfected into insect cells for production of recombinant baculovirus particles.

• Easy colony screening—The parent bacmid in DH10Bac E. coli contains a segment of the lacZa gene. The lacZa gene is disrupted upon transposition of the expression cassette into the bacmid, allowing for blue/white selection of recombinants for easier identification of recombinant colonies.

• High protein expression—The pFastBac vector uses the strong polyhedrin promoter to generate high levels of expression in a variety of insect cell line such as Sf9, Sf21, and High Five cells.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR), Gentamicin (GmR)
Cloning MethodBlunt TOPO™
Expression MechanismCell-Based Expression
FormatLiquid
Product LineBac-to-Bac, TOPO
Protein TagHis Tag (6x)
Quantity20 reactions
Expression SystemBaculovirus
PromoterPolyhedrin
TypeExpression System
Unit SizeEach
Contents & Storage
Bac-to-Bac N-His TOPO Expression Kit:
• Vector kit for 20 TOPO cloning reactions, store at -20°C
   –pFastBac/NT-TOPO vector containing N-terminal His-tag and TEV cleavage site
   –pFastBac 1-Gus control plasmid (control expression vector)
   –Other kit reagents: 10X PCR, dNTP mix, Salt solution, sterile water, control PCR template, polyhedrin forward primer, SV40 pA reverse primer
• One Shot Mach1-T1R Chemically Competent E. coli for 20 reactions (20 x 50 μL), store at -80°C
• MAX Efficiency DH10Bac Competent E. coli for 20 reactions (4 kits with 5 x 0.1 mL each), store at -80°C
• ExpiFectamine Sf Transfection Reagent (1 mL), store at 4°C, do not freeze

Frequently asked questions (FAQs)

Which strand is produced using M13K07 to make ssDNA from pPROEX HT or pFastBAC vectors?

You will get the antisense strand with pProEXHT.
You will get the + or sense strand with pFastBac I of pFastBac HT.

Citations & References (1)

Citations & References
Abstract
Role of Nrf2 and protective effects of Metformin against tobacco smoke-induced cerebrovascular toxicity.
Authors:Prasad S, Sajja RK, Kaisar MA, Park JH, Villalba H, Liles T, Abbruscato T, Cucullo L
Journal:Redox Biol
PubMed ID:28212524
Cigarette smoking (CS) is associated with vascular endothelial dysfunction in a causative way primarily related to the TS content of reactive oxygen species (ROS), nicotine, and inflammation. TS promotes glucose intolerance and increases the risk of developing type-2 diabetes mellitus (2DM) with which it shares other pathogenic traits including the ... More