Tryptamine, 98+%
Tryptamine, 98+%
Tryptamine, 98+%
Thermo Scientific Chemicals

Tryptamine, 98+%

CAS: 61-54-1 | C10H12N2 | 160.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11116.09
also known as A11116-09
Price (USD)
49.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
49.00
Each
Chemical Identifiers
CAS61-54-1
IUPAC Name2-(1H-indol-3-yl)ethan-1-amine
Molecular FormulaC10H12N2
InChI KeyAPJYDQYYACXCRM-UHFFFAOYSA-N
SMILESNCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2.0%
Melting Point (clear melt)109.0-119.0°C
View more

Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1

  • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. J. H. GADDUM,; Z. P. PICARELLI.TWO KINDS OF TRYPTAMINE RECEPTOR . British Journal of Pharmacology. 1997, 120 (S1),134-139.
  2. Gaddum J. H. Tryptamine receptors. The Journal of Physiology. 1953, 119(2-3),363-368.