4-Bromoveratrole, 97%
4-Bromoveratrole, 97%
4-Bromoveratrole, 97%
Thermo Scientific™

4-Bromoveratrole, 97%

CAS: 2859-78-1 | C8H9BrO2 | 217.062 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11133.22
also known as A11133-22
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS2859-78-1
IUPAC Name4-bromo-1,2-dimethoxybenzene
Molecular FormulaC8H9BrO2
InChI KeyKBTMGSMZIKLAHN-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5705-1.5755 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

Solubility
Soluble in benzene, ether, toluene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Han Yu Xin.; Edward R. Biehl. Reaction of various nucleophiles with 2-bromo-p-xylene and 4-bromoveratrole via aryne reaction. J. Org. Chem. 1983, 48, (23), 4397-4399.
  2. Timothy K. Beng.; Robert E. Gawley. Application of Catalytic Dynamic Resolution of N-Boc-2-lithiopiperidine to the Asymmetric Synthesis of 2-Aryl and 2-Vinyl Piperidines. Org. Lett. 1983, 13, (3), 394-397.
  3. In the presence of sodamide, displacement by nucleophiles, e.g. anions of secondary amines or nitriles, occurs by an ortho-metallation-benzyne mechanism. The incoming nucleophile adds regioselectively such that the resulting anion is ortho-stabilized by methoxyl: J. Org. Chem., 48, 4397 (1983):
  4. The derived Grignard reagent has been used in the synthesis of the isoquinoline alkaloid (-)-mesembrine: J. Org. Chem., 60, 6785 (1995).