Blasticidin S HCl (10 mg/mL)
Gibco™

Blasticidin S HCl (10 mg/mL)

Blasticidin S is a peptidyl nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis inRead more
Catalog NumberQuantity
A111390220 mL
A111390310 x 1 mL
Catalog number A1113902
Price (JPY)
167,200
Each
Quantity:
20 mL
Blasticidin S is a peptidyl nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis in in bacteria and eukaryotes, while also active against fungi, nematodes, and tumor cells. Blasticidin S acts by blocking hydrolysis of peptidyl-tRNA induced by release factors and inhibits peptide bond formation. It is used as a selection agent in both mammalian cells and bacterial cells. The recommended working concentration ranges from 1 to 30 μg/mL depending on the cell line and 25–100 μg/mL for bacterial selection. Cell death occurs rapidly, and blasticidin-resistant stable mammalian cell lines can be generated in less than one week.

Resistance to blasticidin S is conferred by BSR and BSD, isolated from Bacillus cereus K55-S and Aspergillus terreus respectively. The BSR resistance gene encodes blasticidin S deaminase, which catalyzes the conversion of blasticidin S to deaminohydroxyblasticidin S. Deaminohydroxyblasticidin S is a biologically inactive derivative of blasticidin S and does not interact with or inhibit prokaryotic or eukaryotic ribosomes. The BSD resistance gene also encodes a blasticidin S deaminase, which catalyzes a similar reaction to the BSR deaminase. For bacterial selection purposes, the salt content of the LB medium must remain low (less than 90 mM) and the pH should not exceed 7.0 to maintain the activity of basticidin S. A kill curve is recommended in order to determine the minimum effective blasticidin S concentration to kill non-resistant cells.

Applications
View detailed protocols on blasticidin selection in mammalian cells, E. coli, and yeast.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEukaryotic Cells, Prokaryotic Cells
Concentration10 mg/mL
Culture TypeMammalian Cell Culture, Insect Cell Culture
Product LineGibco
Quantity20 mL
Shelf Life9 Months
FormLiquid
Product TypeAntibiotic
SterilitySterile-filtered
With AdditivesHEPES
Unit SizeEach
Contents & Storage
Storage conditions: -5°C to -20°C (protect from light)
Shipping conditions: Dry ice
Shelf life: 9 months from date of manufacture

Frequently asked questions (FAQs)

Which of your antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for stable selection in mammalian cells?

All of our antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for making multiple stable cell lines. However, kill curves will need to be performed for each combination of antibiotics since sensitivity to a given antibiotic tends to increase when combined with other antibiotics.

What are the recommended concentrations of antibiotics to use for selection in prokaryotes and eukaryotes?

For best results, optimal concentrations for selection should be determined empirically in each unique experiment through dose response curves. However, to get a general idea of concentrations that have worked for individual cell types, please click on the following url: http://www.thermofisher.com/us/en/home/life-science/cell-culture/transfection/selection.html or type in “Selection Antibiotics” into our main search on www.thermofisher.com.

What is the mode of action on the following antibiotics: Blasticidin, Geneticin (G418), Hygromycin, and Zeocin?

Blasticidin: Nucleoside Inhibits protein synthesis in prokaryotic and eukaryotic cells by interfering with peptidyl transfer reaction of protein synthesis, causing early termination of translation.

Geneticin (G418): Aminoglycoside Blocks protein synthesis in mammalian cells by interfering with ribosomal function.

Hygromycin: Aminocyclitol Inhibits protein synthesis by disrupting translocation and promoting mistranslation.

Zeocin: Intercalates with DNA and cleaves it.

What is the optimal pH of low salt LB for LB + blasticidin plates?

We recommend a pH of 7 or less and half the normal amount of NaCl in your LB media or plates.

See the following paper for details on optimal conditions: Yamaguchi et al (1965) Inhibition of Protein Synthesis by Blasticidin S. Journal of Biochemistry (Tokyo) Volume 57: pp 667-677.

How long can Blasticidin be stored at 4 degrees C after thawing? Does the unused portion have to be discarded after thawing?

Blasticidin is stable for 6 months when stored at 4 degrees C. Discard remaining material after this time.

Citations & References (5)

Citations & References
Abstract
A Scalable, Multiplexed Assay for Decoding GPCR-Ligand Interactions with RNA Sequencing.
Authors:Jones EM, Jajoo R, Cancilla D, Lubock NB, Wang J, Satyadi M, Cheung R, de March C, Bloom JS, Matsunami H, Kosuri S
Journal:Cell Syst
PubMed ID:30904378
'G protein-coupled receptors (GPCRs) are central to how mammalian cells sense and respond to chemicals. Mammalian olfactory receptors (ORs), the largest family of GPCRs, mediate the sense of smell through activation by small molecules, though for most bonafide ligands, they have not been identified. Here, we introduce a platform to ... More
A Brain Penetrant Mutant IDH1 Inhibitor Provides In Vivo Survival Benefit.
Authors:Kopinja J, Sevilla RS, Levitan D, Dai D, Vanko A, Spooner E, Ware C, Forget R, Hu K, Kral A, Spacciapoli P, Kennan R, Jayaraman L, Pucci V, Perera S, Zhang W, Fischer C, Lam MH
Journal:Sci Rep
PubMed ID:29062039
'Mutations in IDH1 are highly prevalent in human glioma. First line treatment is radiotherapy, which many patients often forego to avoid treatment-associated morbidities. The high prevalence of IDH1 mutations in glioma highlights the need for brain-penetrant IDH1 mutant-selective inhibitors as an alternative therapeutic option. Here, we have explored the utility ... More
Biological Characterization of a Stable Effector Functionless (SEFL) Monoclonal Antibody Scaffold in Vitro.
Authors:Liu L, Jacobsen FW, Everds N, Zhuang Y, Yu YB, Li N, Clark D, Nguyen MP, Fort M, Narayanan P, Kim K, Stevenson R, Narhi L, Gunasekaran K, Bussiere JL
Journal:J Biol Chem
PubMed ID:27994063
The stable effector functionLess (SEFL) antibody was designed as an IgG1 antibody with a constant region that lacks the ability to interact with Fc? receptors. The engineering and stability and pharmacokinetic assessments of the SEFL scaffold is described in the accompanying article (Jacobsen, F. W., Stevenson, R., Li, C., Salimi-Moosavi, ... More
Proteomics reveals Rictor as a noncanonical TGF-ß signaling target during aneurysm progression in Marfan mice.
Authors:Parker SJ, Stotland A, MacFarlane E, Wilson N, Orosco A, Venkatraman V, Madrid K, Gottlieb R, Dietz HC, Van Eyk JE
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:30004239
The objective of the present study was to 1) analyze the ascending aortic proteome within a mouse model of Marfan syndrome (MFS; Fbn1
Ubiquitination of DNA Damage-Stalled RNAPII Promotes Transcription-Coupled Repair.
Authors:Nakazawa Y, Hara Y, Oka Y, Komine O, van den Heuvel D, Guo C, Daigaku Y, Isono M, He Y, Shimada M, Kato K, Jia N, Hashimoto S, Kotani Y, Miyoshi Y, Tanaka M, Sobue A, Mitsutake N, Suganami T, Masuda A, Ohno K, Nakada S, Mashimo T, Yamanaka K, Luijsterburg MS, Ogi T
Journal:Cell
PubMed ID:32142649
Transcription-coupled nucleotide excision repair (TC-NER) is initiated by the stalling of elongating RNA polymerase II (RNAPIIo) at DNA lesions. The ubiquitination of RNAPIIo in response to DNA damage is an evolutionarily conserved event, but its function in mammals is unknown. Here, we identified a single DNA damage-induced ubiquitination site in ... More