1-Octene, 97+%
1-Octene, 97+%
1-Octene, 97+%
Thermo Scientific Chemicals

1-Octene, 97+%

CAS: 111-66-0 | C8H16 | 112.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A11146.AP
also known as A11146-AP
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Chemical Identifiers
CAS111-66-0
IUPAC Nameoct-1-ene
Molecular FormulaC8H16
InChI KeyKWKAKUADMBZCLK-UHFFFAOYSA-N
SMILESCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4060-1.4110 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.0%
1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

Solubility
Miscible with water, ether, alcohol and acetone.

Notes
Incompatible with bromine, chlorine, acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Li, Y. Q.; Wang, P.; Zhang, H.; Zhao, X. L.; Lu, Y.; Popović, Z.; Liu, Y. Influence of electrostatic repulsive force and electron-withdrawing effect in ionic diphosphine on regioselectivity of rhodium-catalyzed hydroformylation of 1-octene. J. Mol. Catal. A: Chem. 2015, 402, 37-45.
  2. De Rosa, C.; Tarallo, O.; Auriemma, F.; de Ballesteros, O. R.; Di Girolamo, R.; Malafronte, A. Crystallization behavior and mechanical properties of copolymers of isotactic poly(1-butene) with 1-octene from metallocene catalysts. Polymer 2015, 73, 156-169.