Octanoic acid, 98+%
Octanoic acid, 98+%
Octanoic acid, 98+%
Thermo Scientific Chemicals

Octanoic acid, 98+%

CAS: 124-07-2 | C8H16O2 | 144.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
25 mL
500 mL
2500 mL
Catalog number A11149.0D
also known as A11149-0D
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS124-07-2
IUPAC Nameoctanoic acid
Molecular FormulaC8H16O2
InChI KeyWWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILESCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (Silylated GC)≥98.0% (UK Sourced Material)
FormLiquid
CommentMaterial Sourced in UK and US
Refractive Index1.4260-1.4300 @ 20?C (UK Sourced Material)
View more
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Mizrahi, M.; Almogy, G.; Adar, T.; Lysy, J. Erratum: Dumping syndrome following nissen fundoplication in an adult patient diagnosed by continuous online 13C/12C monitoring of 13C-Octanoic acid breath test a case report. BMC Gastroenterol. 2015, 15 (1), 57.
  2. Royce, L. A.; Yoon, J. M.; Chen, Y.; Rickenbach, E.; Shanks, J. V.; Jarboe, L. R. Evolution for exogenous octanoic acid tolerance improves carboxylic acid production and membrane integrity. Metab. Eng. 2015, 29, 180-188.