3-Iodophenol, 98%
3-Iodophenol, 98%
3-Iodophenol, 98%
Thermo Scientific Chemicals

3-Iodophenol, 98%

CAS: 626-02-8 | C6H5IO | 220.009 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A11155.18
also known as A11155-18
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS626-02-8
IUPAC Name3-iodophenol
Molecular FormulaC6H5IO
InChI KeyFXTKWBZFNQHAAO-UHFFFAOYSA-N
SMILESOC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or fused solid
Melting Point (clear melt)36.0-42.0?C
Appearance (Color)White to pale cream to cream to brown to pale gray
Assay (GC)≥97.5%
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only