Hemin (porcine), 97+%
Hemin (porcine), 97+%
Hemin (porcine), 97+%
Thermo Scientific Chemicals

Hemin (porcine), 97+%

CAS: 16009-13-5 | C34H32FeN4O4 | 616.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11165.06
also known as A11165-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS16009-13-5
IUPAC Name3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
Molecular FormulaC34H32ClFeN4O4
InChI KeyBTIJJDXEELBZFS-HXFTUNQESA-K
SMILESCC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple or brown to black
CommentMaterial Sourced in UK and US
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms (UK Sourced Material)
Assay (unspecified)≥97.0%
View more
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

Solubility
Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Lu, C. H.; Guo, W.; Qi, X. J.; Neubauer, A.; Paltiel, Y.; Willner, I. Hemin-G-quadruplex-crosslinked poly-N-isopropylacrylamide hydrogel: a catalytic matrix for the deposition of conductive polyaniline. Chem. Sci. 2015, 6 (11), 6659-6664.
  2. Preda, L.; Negrila, C.; Lazarescu, M. F.; Anastasescu, M.; Dobrescu, G.; Santos, E.; Lazarescu, V. Hemin interaction with bare and 4,4'-thio-bis-benzene-thiolate covered n-GaAs (110) electrodes. Phys. Chem. Chem. Phys. 2011, 13 (38), 17104-17114.