Platinum, 5% on carbon, dry
Platinum, 5% on carbon, dry
Platinum, 5% on carbon, dry
Thermo Scientific Chemicals

Platinum, 5% on carbon, dry

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11186.03
also known as A11186-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Assay (unspecified)Platinum: 4.4 - 5.5%
FormPowder
Residual water=< 5.0%
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts). Insoluble in water, mineral acids, and organic acids.

Notes
Do not store together with acids. Incompatible with Strong acids, strong oxidizing agents & organics Solvent vapors. Store under inert gas.
RUO – Research Use Only

General References:

  1. Irving Langmuir. The absorption of gases on plane surfaces of glass, mica and platinum. J. Am. Chem. Soc. 1918, 40 (9), 1361-1403.
  2. Platinum catalysts have a much greater tendency than palladium to reduce aromatic and heteroaromatic rings, and a reduced tendency to bring about hydrogenolysis e.g. of benzyl groups. Other applications of platinum catalysts include: reduction of double bonds; aromatic nitro compounds to amines, and to N-arylhydroxylamines in alcoholic solution, containing 1-2% DMSO; hydrazones to hydrazines; imines to amines; benzonitriles to dibenzylamines in the presence of 0.5 mole water.
  3. For a monograph on catalytic hydrogenation, see: P. N. Rylander, Hydrogenation Methods, Academic Press, N.Y. (1985).