2-Mercapto-5-methylbenzimidazole, 98%
2-Mercapto-5-methylbenzimidazole, 98%
2-Mercapto-5-methylbenzimidazole, 98%
Thermo Scientific Chemicals

2-Mercapto-5-methylbenzimidazole, 98%

CAS: 27231-36-3 | C8H8N2S | 164.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11192.06
also known as A11192-06
Price (USD)
23.65
Online exclusive
25.90
Save 2.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.65
Online exclusive
25.90
Save 2.25
Each
Chemical Identifiers
CAS27231-36-3
IUPAC Name5-methyl-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular FormulaC8H8N2S
InChI KeyCWIYBOJLSWJGKV-UHFFFAOYSA-N
SMILESCC1=CC=C2NC(=S)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream to cream to pale brown
Assay (HPLC)≥97.5%
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Kyung Soo Pyo.; Chung Kun Song. The effects of simultaneous treatment of SiO2 gate and Au electrode with octadecyltrichlorosilane and charge transfer molecules on characteristics of pentacene thin film transistors. Thin Solid Films. 2005, 485, (1-2), 230-234.
  2. Bo-Lennart Johansson.; Bjorn Persson. Polarographic study of alkyl benzimidazolyl sulfoxide and the corresponding sulfide and sulfones. Analytica Chimica Acta. 1978, 102, 121-131.