2-Aminonicotinic acid, 98+%
2-Aminonicotinic acid, 98+%
2-Aminonicotinic acid, 98+%
Thermo Scientific Chemicals

2-Aminonicotinic acid, 98+%

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11195.14
also known as A11195-14
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS5345-47-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0% (non-U.S. specification)
Appearance (Color)White to cream to yellow to pale brown
FormPowder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (HPLC: U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only