2,3-Dichlorotoluene, 98%
2,3-Dichlorotoluene, 98%
2,3-Dichlorotoluene, 98%
Thermo Scientific Chemicals

2,3-Dichlorotoluene, 98%

CAS: 32768-54-0 | C7H6Cl2 | 161.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
Catalog number A11206.18
also known as A11206-18
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS32768-54-0
IUPAC Name1,2-dichloro-3-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyGWLKCPXYBLCEKC-UHFFFAOYSA-N
SMILESCC1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.5495-1.5535 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only