3-Chloro-4-fluorobenzenesulfonyl chloride, 98%
3-Chloro-4-fluorobenzenesulfonyl chloride, 98%
3-Chloro-4-fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

3-Chloro-4-fluorobenzenesulfonyl chloride, 98%

CAS: 91170-93-3 | C6H3Cl2FO2S | 229.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11222.14
also known as A11222-14
Price (USD)
77.65
Online Exclusive
85.70
Save 8.05 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
85.70
Save 8.05 (9%)
Each
Chemical Identifiers
CAS91170-93-3
IUPAC Name3-chloro-4-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3Cl2FO2S
InChI KeyDXFXNSNBZNELII-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5500-1.5540 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only