4,4'-Difluorobenzhydryl chloride, 98%
4,4'-Difluorobenzhydryl chloride, 98%
4,4'-Difluorobenzhydryl chloride, 98%
Thermo Scientific Chemicals

4,4'-Difluorobenzhydryl chloride, 98%

CAS: 27064-94-4 | C13H9ClF2 | 238.662 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number A11226.06
also known as A11226-06
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.40
Save 5.75 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 30-Jun-2026
37.40
Save 5.75 (15%)
Each
Chemical Identifiers
CAS27064-94-4
IUPAC Name1-[chloro(4-fluorophenyl)methyl]-4-fluorobenzene
Molecular FormulaC13H9ClF2
InChI KeyFHPNLCLHMNPLEW-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(Cl)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5550-1.5590 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only