D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
Thermo Scientific Chemicals

D-(-)-Tartaric acid, 99%

CAS: 147-71-7 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11264.14
also known as A11264-14
Price (USD)
43.50
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
43.50
Each
Chemical Identifiers
CAS147-71-7
IUPAC Name2,3-dihydroxybutanedioic acid
Molecular FormulaC4H6O6
InChI KeyFEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)163.0-172.0?C (UK-sourced material)
Optical Rotation-13.5 ± 0.5? (C=10 in water, UK-sourced material)
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (UK-sourced material)
View more
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Mahapatra, M.; Tysoe, W. T. Structure and decomposition pathways of D-(-)-tartaric acid on Pd(111). Surface Science 2014, 629, 132-138.
  2. Hu, G.; Zhang, H.; Miao, H.; Wang, J.; Xu, Y. Synthesis, structures and properties of two new chiral rare earth-organic frameworks constructed by l/d-tartaric acid. J. Solid State Chem. 2015, 229, 208-212.