2,4,6-Triisopropylbenzoyl chloride, 98+%
2,4,6-Triisopropylbenzoyl chloride, 98+%
2,4,6-Triisopropylbenzoyl chloride, 98+%
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzoyl chloride, 98+%

CAS: 57199-00-5 | C16H23ClO | 266.809 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11280.06
also known as A11280-06
Price (USD)
61.65
Online Exclusive
68.10
Save 6.45 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.10
Save 6.45 (9%)
Each
Chemical Identifiers
CAS57199-00-5
IUPAC Name2,4,6-tris(propan-2-yl)benzoyl chloride
Molecular FormulaC16H23ClO
InChI KeyOSKNTKJPGKHDHV-UHFFFAOYSA-N
SMILESCC(C)C1=CC(C(C)C)=C(C(Cl)=O)C(=C1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or crystalline powder or powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Identification (FTIR)Conforms
View more
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
RUO – Research Use Only