2-Acetyl-5-methylthiophene, 98%
2-Acetyl-5-methylthiophene, 98%
2-Acetyl-5-methylthiophene, 98%
Thermo Scientific Chemicals

2-Acetyl-5-methylthiophene, 98%

CAS: 13679-74-8 | C7H8OS | 140.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11282.06
also known as A11282-06
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.20
Save 9.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.20
Save 9.55 (15%)
Each
Chemical Identifiers
CAS13679-74-8
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5590-1.5630 @ 20?C
FormLiquid
Appearance (Color)Clear pale yellow to yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only