2,6-Difluorobenzoyl chloride, 98%
2,6-Difluorobenzoyl chloride, 98%
2,6-Difluorobenzoyl chloride, 98%
Thermo Scientific Chemicals

2,6-Difluorobenzoyl chloride, 98%

CAS: 18063-02-0 | C7H3ClF2O | 176.547 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11286.22
also known as A11286-22
Price (USD)
208.65
Online Exclusive
232.00
Save 23.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
208.65
Online Exclusive
232.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS18063-02-0
IUPAC Name2,6-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyQRHUZEVERIHEPT-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4970-1.5015 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only