2-Methylhexanoic acid, 98%
2-Methylhexanoic acid, 98%
2-Methylhexanoic acid, 98%
Thermo Scientific Chemicals

2-Methylhexanoic acid, 98%

CAS: 4536-23-6 | C7H13O2 | 129.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11290.06
also known as A11290-06
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.50
Save 7.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.50
Save 7.85
Each
Chemical Identifiers
CAS4536-23-6
IUPAC Name(2R)-2-methylhexanoate
Molecular FormulaC7H13O2
InChI KeyCVKMFSAVYPAZTQ-ZCFIWIBFSA-M
SMILESCCCC[C@@H](C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.4185-1.4225 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only