Melamine, 99%
Melamine, 99%
Melamine, 99%
Thermo Scientific Chemicals

Melamine, 99%

CAS: 108-78-1 | C3H6N6 | 126.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A11295.0E
also known as A11295-0E
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.80
Save 13.15 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.80
Save 13.15 (15%)
Each
Chemical Identifiers
CAS108-78-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.

Solubility
Slightly soluble in water, hot alcohol, glycerol, glycol and pyridine. Insoluble in ether, benzene and carbon tetrachloride.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Liu, M.; Liu, B.; Shi, L.; Wang, F.; Liang, L.; Sun, J. Melamine-ZnI2 as heterogeneous catalysts for efficient chemical fixation of carbon dioxide to cyclic carbonates. RSC Adv. 2015, 5 (2), 960-966.
  2. Xin, J. Y.; Zhang, L. X.; Chen, D. D.; Lin, K.; Fan, H. C.; Wang,Y.; Xia, C. G. Colorimetric detection of melamine based on methanobactin-mediated synthesis of gold nanoparticles. Food Chem. 2015, 174, 473-479.