3-Methyl-4-nitrobenzonitrile, 97%
3-Methyl-4-nitrobenzonitrile, 97%
3-Methyl-4-nitrobenzonitrile, 97%
Thermo Scientific Chemicals

3-Methyl-4-nitrobenzonitrile, 97%

CAS: 96784-54-2 | C8H6N2O2 | 162.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number A11301.14
also known as A11301-14
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS96784-54-2
IUPAC Name3-methyl-4-nitrobenzonitrile
Molecular FormulaC8H6N2O2
InChI KeyIHVNKSXTJZNBQA-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)72.5-81.5°C
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

Solubility
Insoluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only