Paraformaldehyde, 96%
Paraformaldehyde, 96%
Paraformaldehyde, 96%
Thermo Scientific Chemicals

Paraformaldehyde, 96%

CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
100 g
500 g
2500 g
Catalog number A11313.0C
also known as A11313-0C
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
313.00
Save 46.35 (15%)
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
313.00
Save 46.35 (15%)
Each
Chemical Identifiers
CAS30525-89-4
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or granules or beads
Assay≥95.0 to ≤105.0%
Identification (FTIR)Conforms
Paraformaldehyde is used as a fumigant, a disinfectant and in the manufacture of synthetic resins like melamine resin, phenol resin and polyacetal resin. It is also used in cell culture. It also serves as a fixative in electron microscopy. It is also used in the preparation of formalin fixatives for tissues or cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only