Sodium dihydrogen phosphate dihydrate, 98+%
Sodium dihydrogen phosphate dihydrate, 98+%
Sodium dihydrogen phosphate dihydrate, 98+%
Thermo Scientific Chemicals

Sodium dihydrogen phosphate dihydrate, 98+%

CAS: 13472-35-0 | H6NaO6P | 156.006 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11316.36
also known as A11316-36
Price (USD)
49.50
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
49.50
Each
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
RUO – Research Use Only
L Bohaty; S Haussühl. Elastic, electrostrictive, and electro-optic properties of orthorhombic sodium dihydrogen phosphate dihydrate. Zeitschrift für Kristallographie-Crystalline Materials.1984, 167, 303-306.