3,5-Dimethylphenylhydrazine hydrochloride, 97%
3,5-Dimethylphenylhydrazine hydrochloride, 97%
3,5-Dimethylphenylhydrazine hydrochloride, 97%
3,5-Dimethylphenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

3,5-Dimethylphenylhydrazine hydrochloride, 97%

CAS: 60481-36-9 | C8H13ClN2 | 172.656 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11336.06
also known as A11336-06
Price (USD)
42.65
Online Exclusive
46.80
Save 4.15 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
46.80
Save 4.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS60481-36-9
IUPAC Name(3,5-dimethylphenyl)hydrazine hydrochloride
Molecular FormulaC8H13ClN2
InChI KeyRSBQANBNDXZFIY-UHFFFAOYSA-N
SMILESCl.CC1=CC(NN)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder or lumps
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Oluseye K. Onajole,; Marco Pieroni,; Suresh K. Tipparaju,; Shichun Lun,; Jozef Stec,; Gang Chen,; Hendra Gunosewoyo,; Haidan Guo,; Nicole C. Ammerman,;, William R. Bishai,; Alan P. Kozikowski. Preliminary Structure-Activity Relationships and Biological Evaluation of Novel Antitubercular Indolecarboxamide Derivatives Against Drug-Susceptible and Drug-Resistant Mycobacterium tuberculosis Strains . J. Med. Chem.,. 2013, 56 (10), 4093-4103.