1-Bromodecane, 98%
1-Bromodecane, 98%
1-Bromodecane, 98%
Thermo Scientific Chemicals

1-Bromodecane, 98%

CAS: 112-29-8 | C10H21Br | 221.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
100 g
500 g
2500 g
Catalog number A11340.22
also known as A11340-22
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.30
Save 4.65 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.30
Save 4.65 (15%)
Each
Chemical Identifiers
CAS112-29-8
IUPAC Name1-bromodecane
Molecular FormulaC10H21Br
InChI KeyMYMSJFSOOQERIO-UHFFFAOYSA-N
SMILESCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to pale yellow
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.4540-1.4580 @ 20?C
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only
Gary M. McNamee; Brian C. Willett; Daniel M. La Perriere; Dennis G. Peters. Electrochemical reduction of 1-iododecane and 1-bromodecane at a mercury cathode in dimethylformamide. J. Am. Chem. Soc.1977, 99, (6)1831-1835.