2-Benzothiazoleacetonitrile, 98%
2-Benzothiazoleacetonitrile, 98%
2-Benzothiazoleacetonitrile, 98%
2-Benzothiazoleacetonitrile, 98%
Thermo Scientific Chemicals

2-Benzothiazoleacetonitrile, 98%

CAS: 56278-50-3 | C9H6N2S | 174.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11343.14
also known as A11343-14
Price (USD)
300.65
Online Exclusive
334.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
300.65
Online Exclusive
334.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS56278-50-3
IUPAC Name2-(1,3-benzothiazol-2-yl)acetonitrile
Molecular FormulaC9H6N2S
InChI KeyZMZSYUSDGRJZNT-UHFFFAOYSA-N
SMILESN#CCC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow or pale cream to brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)99.5-105.5?C
2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents .
RUO – Research Use Only
Ram Vishnu Ji; Srivastava Pratibha; Agarwal Nidhi. Carbanion induced synthesis of pyrido [2, 1-b] benzothiazoles through ring transformation reactions.Journal of Chemical Research.2001, 2001 (10), 421-423.