Butyrophenone, 99%
Butyrophenone, 99%
Butyrophenone, 99%
Thermo Scientific Chemicals

Butyrophenone, 99%

CAS: 495-40-9 | C10H12O | 148.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Catalog number A11345.14
also known as A11345-14
Price (USD)
37.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
37.60
Each
Chemical Identifiers
CAS495-40-9
IUPAC Name1-phenylbutan-1-one
Molecular FormulaC10H12O
InChI KeyFFSAXUULYPJSKH-UHFFFAOYSA-N
SMILESCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
RUO – Research Use Only

General References:

  1. Paul A. J. Janssen; Anton H. M. Jageneau; Karel H. L. Schellekens. Chemistry and pharmacology of compounds related to 4-(4-hydroxy-4-phenyl-piperidino)-butyrophenone.Psychopharmacology.1960, 1, 389-392.
  2. Karl Ekbom; Halvar Lindholm; Lennart Ljungberg. New dystonic syndrome associated with butyrophenone therapy.Original Investigations.1972, 202, 94-103.