5-Chlorothiophene-2-sulfonyl chloride, 97%
5-Chlorothiophene-2-sulfonyl chloride, 97%
5-Chlorothiophene-2-sulfonyl chloride, 97%
Thermo Scientific Chemicals

5-Chlorothiophene-2-sulfonyl chloride, 97%

CAS: 2766-74-7 | C4H2Cl2O2S2 | 217.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number A11348.03
also known as A11348-03
Price (USD)
17.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
17.10
Each
Chemical Identifiers
CAS2766-74-7
IUPAC Name5-chlorothiophene-2-sulfonyl chloride
Molecular FormulaC4H2Cl2O2S2
InChI KeySORSTNOXGOXWAO-UHFFFAOYSA-N
SMILESClC1=CC=C(S1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5855-1.5905 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow to yellow to brown
Assay (GC)≥96.0%
5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
RUO – Research Use Only
Varun Rawat; Pandurang V. Chouthaiwale; Vilas B. Chavan; Gurunath Suryavanshi; Arumugam Sudalai. A facile enantioselective synthesis of (S)-N-(5-chlorothiophene-2-sulfonyl)-β,β-diethylalaninol via proline-catalyzed asymmetric α-aminooxylation and α-amination of aldehyde.Tetrahedron Letters.2010, 51, 6565-6567.