2-Chloro-3-hydroxypyridine, 98%
2-Chloro-3-hydroxypyridine, 98%
2-Chloro-3-hydroxypyridine, 98%
Thermo Scientific Chemicals

2-Chloro-3-hydroxypyridine, 98%

CAS: 6636-78-8 | C5H4ClNO | 129.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11376.14
also known as A11376-14
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS6636-78-8
IUPAC Name2-chloropyridin-3-ol
Molecular FormulaC5H4ClNO
InChI KeyRSOPTYAZDFSMTN-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)168.0-174.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only