Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A11379.18
also known as A11379-18
Price (USD)
38.80
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
38.80
Each
Chemical Identifiers
CAS1185-53-1
IUPAC Namehydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular FormulaC4H12ClNO3
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥99.0% to ≤101.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Biological buffer.
  2. Lin, L.; Pan, Q. ZnFe2O4@ C/graphene nanocomposites as excellent anode materials for lithium batteries. J. Mater. Chem. A 2015, 3 (4), 1724-1729.
  3. Oishi, M. Enzyme-free and isothermal detection of microRNA based on click-chemical ligation-assisted hybridization coupled with hybridization chain reaction signal amplification. Anal. Bioanal. Chem. 2015, 407 (14), 4165-4172.