p-Tolualdehyde, 98%
p-Tolualdehyde, 98%
p-Tolualdehyde, 98%
Thermo Scientific Chemicals

p-Tolualdehyde, 98%

CAS: 104-87-0 | C8H8O | 120.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
2500 g
Catalog number A11384.0E
also known as A11384-0E
Price (USD)
556.65
Online Exclusive
618.00
Save 61.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
556.65
Online Exclusive
618.00
Save 61.35 (10%)
Each
Chemical Identifiers
CAS104-87-0
IUPAC Name4-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyFXLOVSHXALFLKQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Free acid (titration)≤2.5%
Identification (FTIR)Conforms
View more
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
RUO – Research Use Only

General References:

  1. Jose A. Cabello.; Juan M. Campelo.; Angel Garcia.; Diego Luna.; Jose M. Marinas. Knoevenagel condensation in the heterogeneous phase using aluminum phosphate-aluminum oxide as a new catalyst. J. Org. Chem. 1984, 49, (26), 5195-5197.
  2. Daniel L. Comins.; Jack D. Brown. Ortho metalation directed by .alpha.-amino alkoxides . J. Org. Chem. 1984, 49, (6), 1078-1083.