3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals
3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals
3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A11389.09
also known as A11389-09
10 g
Catalog number A11389.09
also known as A11389-09
Price (USD)
48.65
Online Exclusive
54.20
Save 5.55 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
54.20
Save 5.55 (10%)
Each
Chemical Identifiers
CAS618-48-4
IUPAC Name3-chlorobenzamide
Molecular FormulaC7H6ClNO
InChI KeyMJTGQALMWUUPQM-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)132.0-138.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only