Benzopinacol, 98%
Benzopinacol, 98%
Benzopinacol, 98%
Thermo Scientific Chemicals

Benzopinacol, 98%

CAS: 464-72-2 | C26H22O2 | 366.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11421.22
also known as A11421-22
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS464-72-2
IUPAC Nametetraphenylethane-1,2-diol
Molecular FormulaC26H22O2
InChI KeyMFEWNFVBWPABCX-UHFFFAOYSA-N
SMILESOC(C1=CC=CC=C1)(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to very pale cream or very pale grey
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications: Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
RUO – Research Use Only