Collagen I, Coated Plate, 6 well - FAQs
Collagen I, Coated Plate, 6 well - FAQs
View additional product information for Collagen I, Coated Plate, 6 well - FAQs (A1142801)
6 product FAQs found
Do I need to use all plates from the Collagen I, Coated Plate, 6 well, (Cat. No. A1142801) package at once or can I store the unused plates? If so, for how long can they be stored?
The unused plates can be resealed in the foil bag and stored for the remaining shelf life of the product.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the dimensions for the Collagen I, Coated Plate, 6 well (Cat. No. A1142801)?
The dimensions for the Collagen I, Coated Plate, 6 well are as follows:
- Height: 14.4 mm
- Length: 127.76 mm
- Width: 85.48 mm
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What can be used to detach cells from plates when coated with a thin layer of collagen I?
Enzymes that can be used include: Gibco TrypLE Express Enzyme, Gibco TrypLE Select Enzyme, or Gibco StemPro Accutase Cell Dissociation Reagent.
Find additional tips, troubleshooting help, and resources within our Extracellular Matrices (ECM) Support Center.
What cell types are most commonly cultured in collagen I?
The following cell types are commonly cultured in collagen I:
- Endothelial cells (including HUVEC)
- Fibroblasts
- Hepatocytes
- Chondrocytes
- Osteoclasts
- Osteoblasts
- Keratinocytes
- Corneal epithelium
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How is collagen I used?
The key applications include the following:
- Collagen I induces microvascular endothelial cells to adopt a spindle-shaped morphology, in vitro, and to align into solid cord-like assemblies. Vascular endothelial cells can also form vessel-like, tubular structures when cultured on collagen I. Collagen I can be used for in vitro angiogenesis assays.
- Breast cancer stem cells can undergo differentiation when cultured on collagen I.
- Collagen I has been used for the culture of primary colon carcinoma cell lines; mouse liver progenitor cells have been cultured in 3D collagen I and rat pancreatic islets.
- Collagen I can act as a barrier in cell invasion assays, and has been used to study cell adhesion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are some general differences between Gibco Geltrex Matrix, collagen, AlgiMatrix 3D Culture System, and CTS Cellstart Substrate?
Gibco Geltrex Matrix and collagen (rat tail and bovine) can be used as either a coating solution or a 3D gel matrix. CELLstart Substrate was developed to be used as a xeno-free coating matrix for only ESC applications (as a substitute for Gibco Geltrex Matrix or Matrigel Matrix, which are of animal origin). AlgiMatrix Matrix is a 3D scaffold-type matrix that does not support cell attachment but does provide a good environment for growing spheroids that can be easily harvested for downstream applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.