What can be used to detach cells from plates when coated with a thin layer of collagen I?
Enzymes that can be used include: Gibco TrypLE Express Enzyme, Gibco TrypLE Select Enzyme, or Gibco StemPro Accutase Cell Dissociation Reagent.
Find additional tips, troubleshooting help, and resources within our Extracellular Matrices (ECM) Support Center.
Find additional tips, troubleshooting help, and resources within our Extracellular Matrices (ECM) Support Center.