1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%
Thermo Scientific Chemicals

1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%

CAS: 270573-71-2 | C6H6Na2O9S2 | 332.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11439.22
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Chemical Identifiers
CAS270573-71-2
IUPAC Namedisodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate
Molecular FormulaC6H6Na2O9S2
InChI KeyZLRROLLKQDRDPI-UHFFFAOYSA-L
SMILESO.[Na+].[Na+].OC1=CC(=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Sulfated Ash41.05-44.47% (as monohydrate)
Assay (ex Sulphated Ash)≥96.0 to ≤104.0% (as monohydrate)
Appearance (Color)White to cream
Water Content (Karl Fischer Titration)4.4-6.4% (0.8-1.2 waters of hydration)
View more
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

Solubility
It is readily soluble in water.

Notes
Stable under normal conditions. Keep away from strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Kathryn E. Cole.; Ann M. Valentine. Spermidine and Spermine Catalyze the Formation of Nanostructured Titanium Oxide. Biomacromolecules.20078, (5), 1641-1647.
  2. Reagent for Fe and Ti: Helv. Chim. Acta, 34, 528 (1951).