1-Isopropyl-4-piperidone, 99%
1-Isopropyl-4-piperidone, 99%
1-Isopropyl-4-piperidone, 99%
Thermo Scientific Chemicals

1-Isopropyl-4-piperidone, 99%

CAS: 5355-68-0 | C8H15NO | 141.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11480.18
also known as A11480-18
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS5355-68-0
IUPAC Name4-oxo-1-(propan-2-yl)piperidin-1-ium
Molecular FormulaC8H16NO
InChI KeyCCDBCHAQIXKJCG-UHFFFAOYSA-O
SMILESCC(C)[NH+]1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4635 - 1.4675 @20?C
Formliquid
Appearance (Color)Clear, pale yellow to brown
Assay (GC)> 98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only