3-Iodo-4-methylaniline, 98%
3-Iodo-4-methylaniline, 98%
3-Iodo-4-methylaniline, 98%
Thermo Scientific Chemicals

3-Iodo-4-methylaniline, 98%

CAS: 35944-64-0 | C7H8IN | 233.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11485.06
also known as A11485-06
Price (USD)
43.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.30
Each
Chemical Identifiers
CAS35944-64-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to dark brown
FormCrystals or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)36.0-42.0°C
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

General References:

  1. Gonca Özdemir Tarı,; Sümeyye Gümüş,; Erbil Ağar. Crystal structure, spectroscopic studies and quantum mechanical calculations of 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2015, 115 119-127.
  2. authors. title . Journal name. year of publication , volume issue , pg no.