alpha-Chloralose, 98+%, beta anomer ca 20%
alpha-Chloralose, 98+%, beta anomer ca 20%
alpha-Chloralose, 98+%, beta anomer ca 20%
Thermo Scientific Chemicals

alpha-Chloralose, 98+%, beta anomer ca 20%

CAS: 15879-93-3 | C8H11Cl3O6 | 309.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11492.14
also known as A11492-14
Price (USD)
26.50
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
26.50
Each
Chemical Identifiers
CAS15879-93-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≤20% (Beta chloralose)
Optical Rotation+15? to +19? (c= 2 in Ethanol)
Appearance (Color)White to pale yellow
Melting Point (clear melt)174-185?C
FormPowder
View more
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canaden
RUO – Research Use Only

General References:

  1. Holzgrefe HH,; Everitt JM,; Wright EM. Alpha-chloralose as a canine anesthetic. Journal name. 1987, 37(5):587-595.
  2. E. Dale Crider,; Jimmie C. Mc,; Daniel. Alpha-Chloralose Used to Capture Canada Geese. The Journal of Wildlife Management. 1967 , volume 2, 258-264.