2-Chlorophenyl phosphorodichloridate, 98+%
2-Chlorophenyl phosphorodichloridate, 98+%
2-Chlorophenyl phosphorodichloridate, 98+%
Thermo Scientific Chemicals

2-Chlorophenyl phosphorodichloridate, 98+%

CAS: 15074-54-1 | C6H4Cl3O2P | 245.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11505.22
also known as A11505-22
Price (USD)
253.65
Online Exclusive
282.00
Save 28.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
253.65
Online Exclusive
282.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS15074-54-1
IUPAC Name2-chlorophenyl chlorophosphonochloridate
Molecular FormulaC6H4Cl3O2P
InChI KeyVLDPXPPHXDGHEW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1OP(Cl)(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5370-1.5410 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only