(+/-)-3,3-Dimethyl-2-butylamine, 98%
(+/-)-3,3-Dimethyl-2-butylamine, 98%
(+/-)-3,3-Dimethyl-2-butylamine, 98%
Thermo Scientific Chemicals

(+/-)-3,3-Dimethyl-2-butylamine, 98%

CAS: 3850-30-4 | C6H16N | 102.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11530.14
also known as A11530-14
Price (USD)
746.65
Special offer
Online exclusive
Ends: 30-Jun-2026
878.00
Save 131.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
746.65
Special offer
Online exclusive
Ends: 30-Jun-2026
878.00
Save 131.35 (15%)
Each
Chemical Identifiers
CAS3850-30-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4120-1.4160 @ 20°C
(±)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from acid chlorides and acid anhydrides.
RUO – Research Use Only