ACES, 99%
ACES, 99%
ACES, 99%
Thermo Scientific Chemicals

ACES, 99%

CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11553.22
also known as A11553-22
Price (USD)
145.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
145.00
Each
Chemical Identifiers
CAS7365-82-4
IUPAC Name2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular FormulaC4H10N2O4S
InChI KeyDBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILESNC(=O)CNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
ACES is used as a Good's buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

Solubility
Soluble in water(18g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Chuanmin Ruana.; Kefeng Zenga.; Craig A. Grimes. A mass-sensitive pH sensor based on a stimuli-responsive polymer. Analytica Chimica Acta. 2003, 497, (1-2), 123-131.
  2. Zdunek, A.; Rachowicz, W.; Sehlstedt, N. Toward hp-adaptive solution of 3D electromagnetic scattering from cavities. Computers & Mathematics with Applications. 2005, 49, (1), 23-38.
  3. Biological buffer, pK a = 6.9 at 20 ° : Biochemistry, 5, 467 (1966).