p-Toluamide, 98+%
p-Toluamide, 98+%
p-Toluamide, 98+%
Thermo Scientific Chemicals

p-Toluamide, 98+%

CAS: 619-55-6 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11559.14
also known as A11559-14
Price (USD)
36.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
36.00
Each
Chemical Identifiers
CAS619-55-6
IUPAC Name4-methylbenzamide
Molecular FormulaC8H9NO
InChI KeyUHBGYFCCKRAEHA-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)156.0-163.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only