4-Amino-2,6-dichlorophenol, 97%
4-Amino-2,6-dichlorophenol, 97%
4-Amino-2,6-dichlorophenol, 97%
Thermo Scientific Chemicals

4-Amino-2,6-dichlorophenol, 97%

CAS: 5930-28-9 | C6H5Cl2NO | 178.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11568.18
also known as A11568-18
Price (USD)
86.65
Online exclusive
95.80
Save 9.15
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
86.65
Online exclusive
95.80
Save 9.15
Each
Chemical Identifiers
CAS5930-28-9
IUPAC Name4-amino-2,6-dichlorophenol
Molecular FormulaC6H5Cl2NO
InChI KeyKGEXISHTCZHGFT-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown or pale red
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤1.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only