Oxamide, 98+%
Oxamide, 98+%
Oxamide, 98+%
Thermo Scientific Chemicals

Oxamide, 98+%

CAS: 471-46-5 | C2H4N2O2 | 88.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11570.36
also known as A11570-36
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS471-46-5
IUPAC Nameethanediamide
Molecular FormulaC2H4N2O2
InChI KeyYIKSCQDJHCMVMK-UHFFFAOYSA-N
SMILESNC(=O)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental AnalysisCarbon C: 26.73 - 27.82% (Theory 27.28%)
Formpowder
Assay (HPLC)> 98.0%
Elemental AnalysisNitrogen N: 31.17 - 32.45% (Theory 31.81%)
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Yanga, L.; Liub, L.; Olsena, B.A.; Nussbaum, M.A. The determination of oxalic acid, oxamic acid, and oxamide in a drug substance by ion-exclusion chromatography. Journal of Pharmaceutical and Biomedical Analysis. 2000, 22, (3), 487-493.
  2. Isabel Fernandez.; JoseR. Pedro.; Antonio L. Rosello.; Rafael Ruiz.; Xavier Ottenwaelder.; Yves Journaux. Aerobic epoxidation of olefins catalyzed by square-planar nickel(II) complexes of bis-N,N'-disubstituted oxamides and related ligands. Tetrahedron Letters. 1998, 39, (18), 2869-2872.