Cyclopentanemethanol, 98%
Cyclopentanemethanol, 98%
Cyclopentanemethanol, 98%
Thermo Scientific Chemicals

Cyclopentanemethanol, 98%

CAS: 3637-61-4 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11595.22
also known as A11595-22
Price (USD)
923.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,086.00
Save 162.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
923.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,086.00
Save 162.35 (15%)
Each
Chemical Identifiers
CAS3637-61-4
IUPAC Namecyclopentylmethanol
Molecular FormulaC6H12O
InChI KeyISQVBYGGNVVVHB-UHFFFAOYSA-N
SMILESOCC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4560-1.4600 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only