This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.
Solubility
Miscible with water, alcohol, ether
Notes
Miscible with water, alcohol, ether
RUO – Research Use Only
General References:
- Oswal, S.L.; Desai, H.S. Studies of viscosity and excess molar volume of binary mixtures.: 1. Propylamine+1-alkanol mixtures at 303.15 and 313.15 K. Fluid Phase Equilibria. 1998, 149, (1-2), 359-376.
- Gerhard J. Mohr.; Caspar Demuth.; Ursula E. Spichiger-Keller. Application of Chromogenic and Fluorogenic Reactands in the Optical Sensing of Dissolved Aliphatic Amines. Anal. Chem. 1998, 70, (18), 3868-3873.