4-Methyl-2-pentanone, 99%
4-Methyl-2-pentanone, 99%
4-Methyl-2-pentanone, 99%
Thermo Scientific Chemicals

4-Methyl-2-pentanone, 99%

CAS: 108-10-1 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11618.AP
also known as A11618-AP
Price (USD)
33.65
Online exclusive
37.20
Save 3.55
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
33.65
Online exclusive
37.20
Save 3.55
Each
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.3940-1.3970 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N′-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

Solubility
Miscible with ethanol, ether, acetone, benzene and chloroform. Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong bases. Avoid conditions like flames and sparks.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Negadi, L.; Mokbel, I.; Chiali-Baba-Ahmed, N.; Kara-Zaitri, L. Phase equilibrium properties of binary mixtures containing 2, 5-dimethylfuran and furfuryl alcohol or methyl isobutyl ketone at several temperatures. J. Chem. Thermodyn. 2014, 70, 233-238.
  2. Bagabas, A. A.; Mokhtar, M.; Akhmedov, V. M.; Narasimharao, K.; Basahel, S. N.; Al-Rabiah, A. Ru-C-ZnO Composite Catalysts for the Synthesis of Methyl Isobutyl Ketone via Single Step Gas Phase Acetone Self-Condensation. Catal. Lett. 2014, 144 (7), 1278-1288.