Ethyl tetradecanoate, 98%
Ethyl tetradecanoate, 98%
Ethyl tetradecanoate, 98%
Thermo Scientific Chemicals

Ethyl tetradecanoate, 98%

CAS: 124-06-1 | C16H32O2 | 256.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number A11646.AE
also known as A11646-AE
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.70
Save 8.05 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.70
Save 8.05 (15%)
Each
Chemical Identifiers
CAS124-06-1
IUPAC Nameethyl tetradecanoate
Molecular FormulaC16H32O2
InChI KeyMMKRHZKQPFCLLS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4340-1.4380 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Friedrich Martin Wurst et. al. Concentration of fatty acid ethyl esters in hair of alcoholics: comparison to other biological state markers and self reported-ethanol intake. Alcohol and Alcoholism. 2004, 39 (1), 33-38.
  2. V Auwärter et. al. Fatty acid ethyl esters in hair as markers of alcohol consumption. Segmental hair analysis of alcoholics, social drinkers, and teetotalers. Clinical Chemistry. 2001, 47 (12), 2114-2123.